missing translation for 'onlineSavingsMsg'
Learn More

Molecular Sieve Dehydrate with indicator for drying solvents, Solstice

Product Code. 15636740
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15636740 1 kg 1kg
15646740 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15636740 Supplier Honeywell-Fluka Supplier No. 698391KG
 Restricted Item
Estimated Shipment: 09-04-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

BEADS, WITH INDICATOR FOR DRYING SOLVENTS

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Molecular Sieve Dehydrate Fluka with indicator for drying solvents
Quantity 1 kg
CAS 7646-79-9, 1318-02-1
MDL Number MFCD00147630
UN Number UN3077
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.