missing translation for 'onlineSavingsMsg'
Learn More

Molecular sieves, 13X, 1-2mm (0.04-0.08in) dia. pellets

Product Code. 11309139
Change view
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11309139 1 kg 1kg
11319139 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11309139 Supplier Thermo Scientific Alfa Aesar Supplier No. 087954.A1
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 63231-69-6

Molecular sieves is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals. It is preferred for static dehydration in closed liquid or gas systems, e.g., in packaging of drugs, electric components and perishable chemicals; water scavenging in printing and plastics systems and drying saturated hydrocarbon streams. Generally considered a universal drying agent in polar and nonpolar media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63231-69-6
MDL Number MFCD00131613
IUPAC Name Molecular sieves 13X

Specifications

Particle Size 1/16 in.
Sensitivity Hygroscopic
Quantity 1 kg
Physical Form 1/16in Pellets
Solubility Information Insoluble in water.
Chemical Name or Material Molecular sieves, 13X, 1 to 2mm (0.04 to 0.08 in.) dia. pellets

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.