missing translation for 'onlineSavingsMsg'
Learn More

Molecular sieves, 13X, 3-5 mm (0.12-0.20in) beads

Product Code. 11479820
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1kg
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11479820 5000 g 5000g
11322366 250 g 250g
11332366 1000 g 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11479820 Supplier Thermo Scientific Alfa Aesar Supplier No. L06085.0I
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 63231-69-6

Molecular sieves, 13X, 3-5 mm (0.12-0.20in) beads are used for liquefied petroleum gas, natural gas drying, desulfurization, clean. It is used as an adsorbent for gases and liquids. It is used in chemistry laboratory for separating compounds and drying reaction starting materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63231-69-6
MDL Number MFCD00131613
IUPAC Name Molecular sieves 13X

Specifications

Sensitivity Hygroscopic
Quantity 5000 g
Solubility Information Insoluble in water.
Chemical Name or Material Molecular sieves, 13X, 3 to 5mm (0.12 to 0.20 in) beads

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.