missing translation for 'onlineSavingsMsg'
Learn More

Molecular sieves, 3A, 1-2mm (0.04-0.08in) beads

Product Code. 11327405
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1kg
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11327405 1000 g 1kg
11317405 250 g 250g
11429820 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11327405 Supplier Thermo Scientific Alfa Aesar Supplier No. L05335.0B
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 308080-99-1 | AlNaO6Si2 | 202.135 g/mol

Molecular sieves, 3A is used as a desiccant in petroleum and chemical industries. It is useful for the drying of unsaturated hydrocarbons such as ethylene, propylene and butadiene. It is also used in the removal of water from ethanol. It finds application in the drying of certain chemicals namely ethanol, refrigerants and natural gas. It plays an important role in selective adsorption of water in insulated glass (IG) and polyurethane. It has been considered as a general-purpose drying agent in polar and non polar media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 308080-99-1
Molecular Formula AlNaO6Si2
Molecular Weight (g/mol) 202.135
MDL Number MFCD00131613
InChI Key URGAHOPLAPQHLN-UHFFFAOYSA-N
PubChem CID 19758701
SMILES [O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3]

Specifications

Sensitivity Hygroscopic
Quantity 1000 g
Solubility Information Insoluble in cold water,hot water,methanol,diethyl ether,n-octanol and acetone.
IUPAC Name Molecular sieves 3A
Chemical Name or Material Molecular sieves, 3A, 1 to 2mm (0.04 to 0.08 in) beads
What regeneration temperature do you recommend for Molecular sieves, 3A, 1-2mm (0.04-0.08in) beads?

We recommend a regeneration temperature of 120-315 degrees Celsius for the Molecular sieves, 3A, 1-2mm (0.04-0.08in) beads.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.