missing translation for 'onlineSavingsMsg'
Learn More

Molecular sieves 3A, 4 to 8 mesh

Product Code. 10792621 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
5 kg
Packaging:
Glass bottle
Plastic drum
Unit Size:
500g
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10792621 5 kg Plastic drum 5kg
10104220 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10792621 Supplier Thermo Scientific Acros Supplier No. 197240050
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 308080-99-1 | AlNaO6Si2 | 202.135 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Drying agent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 308080-99-1
Molecular Formula AlNaO6Si2
Molecular Weight (g/mol) 202.135
MDL Number MFCD00131613
InChI Key URGAHOPLAPQHLN-UHFFFAOYSA-N
PubChem CID 19758701
IUPAC Name aluminum;sodium;dioxido(oxo)silane
SMILES [O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3]

Specifications

Color Beige
Bulk Density 700 to 800g/L
Packaging Plastic drum
Residual Water 1.5% max. (550°C, 2 hrs.)
Water Adsorption 20% min. (50%rH, 20°C, 24h)
Quantity 5 kg
Physical Form Granules
Solubility Information Solubility in water: insoluble.
Chemical Name or Material Molecular sieves 3A, 4 to 8 mesh

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.