missing translation for 'onlineSavingsMsg'
Learn More

Molecular Sieves, 3A, Powder, TRC

Product Code. 30477374
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477374 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30477374 Supplier Toronto Research Chemicals Supplier No. TRCM48900350G
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Molecular Sieves, 3A, Powder
CAS 308080-99-1
Molecular Formula AlNaO6Si2
Quantity 50 g
Synonym Zeolites, 3A, 3A Zeolites, 3ASDG, Baylith SG 232, HTMS 3A, LEK 2425, MS 3A, Mizuka Sieves 3A, Molecular Sieve 3A, Molsiv 3A, Purmol 3A, Purmol 3STH, S 136, Siliporite 3A, Siliporite NK 30, Siliporite NK 30AP, Sylobead 562CS, Tricat 3A, UOP-AS 5078, Z 3-02, Zeolum A 3
InChI Formula InChI=1S/Al.Na.2O3Si/c;;2*1-4(2)3/q+3;+1;2*-2
SMILES O=[Si]([O-])[O-].O=[Si]([O-])[O-].[Al+3].[Na+]
Molecular Weight (g/mol) 202.14
Formula Weight 201.89
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.