missing translation for 'onlineSavingsMsg'
Learn More

Molecular Sieves, 4 Å, 8 to 12mesh, Solstice

Product Code. 15651850
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15651850 5 kg 5kg
15641850 250 g 250g
15631850 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15651850 Supplier Honeywell-Fluka Supplier No. 2086045KG
 Restricted Item
Estimated Shipment: 23-03-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Beads, 8-12 mesh

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70955-01-0
Molecular Formula C20H25FN2O8
Molecular Weight (g/mol) 440.42
MDL Number MFCD00131613
InChI Key FJUZEZRZKVMAMD-UHFFFAOYNA-N
PubChem CID 73906282
IUPAC Name (2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid
SMILES COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC

Specifications

Mesh Size 8 to 12mesh
Quantity 5 kg
Linear Formula Na12[(AlO2)12(SiO2)12] · xH2O
UN Number NONH for all modes of transport
Chemical Name or Material Molecular Sieves, 4 Å
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.