missing translation for 'onlineSavingsMsg'
Learn More

Molecular Sieves, 5Å, Solstice

Product Code. 15671850
Change view
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Drum with Inliner
Poly Bottle
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15671850 5 kg Drum with Inliner 5kg
15661850 1 kg Poly Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15671850 Supplier Honeywell-Fluka Supplier No. 2086205KG
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Beads, 8-12 mesh

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69912-79-4
Molecular Formula AlNaO6Si2
Molecular Weight (g/mol) 202.14
MDL Number MFCD00147627
InChI Key URGAHOPLAPQHLN-UHFFFAOYSA-N
PubChem CID 19758701
SMILES [Na+].[Al+3].[O-][Si]([O-])=O.[O-][Si]([O-])=O

Specifications

Mesh Size 8 to 12mesh
Packaging Drum with Inliner
Pore Size 5 Å
Quantity 5 kg
Physical Form Beads
Linear Formula Ca/nNa12-2n[(AlO2)12(SiO2)12] · xH2O
UN Number NONH for all modes of transport
IUPAC Name aluminum;sodium;dioxido(oxo)silane
Chemical Name or Material Molecular Sieves, 5Å
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.