missing translation for 'onlineSavingsMsg'
Learn More

Molecular Sieves, 5 Å Pellets, 3.2mm, Solstice

Product Code. 15644110 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15644110 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15644110 Supplier Honeywell-Fluka Supplier No. 3410291KG
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Pellets, 3.2 mm

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Packaging Poly bottle
Quantity 1 kg
CAS 69912-79-4
Molecular Formula AlNaO6Si2
Linear Formula Ca/nNa12-2n[(AlO2)12(SiO2)12] · xH2O
MDL Number MFCD00147627
UN Number NONH for all modes of transport
InChI Key URGAHOPLAPQHLN-UHFFFAOYSA-N
SMILES [Na+].[Al+3].[O-][Si]([O-])=O.[O-][Si]([O-])=O
IUPAC Name aluminum;sodium;dioxido(oxo)silane
Molecular Weight (g/mol) 202.14
PubChem CID 19758701
Chemical Name or Material Molecular sieves, 5Å
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.