missing translation for 'onlineSavingsMsg'
Learn More

Mollugin, TRC

Product Code. 30474456
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30474456 1 g 1g
30467260 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30474456 Supplier Toronto Research Chemicals Supplier No. TRCM4876001G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55481-88-4
Molecular Formula C17 H16 O4
Molecular Weight (g/mol) 284.31
Synonym 6-Hydroxy-2,2-dimethyl-2H-naphtho(1,2-b)pyran-5-carboxylic acid methyl ester, Rubimaillin
IUPAC Name methyl 6-hydroxy-2,2-dimethylbenzo[h]chromene-5-carboxylate
SMILES COC(=O)c1c(O)c2ccccc2c3OC(C)(C)C=Cc13

Specifications

Quantity 1 g
InChI Formula InChI=1S/C17H16O4/c1-17(2)9-8-12-13(16(19)20-3)14(18)10-6-4-5-7-11(10)15(12)21-17/h4-9,18H,1-3H3
Formula Weight 284.1049
Chemical Name or Material Mollugin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.