missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Molnupiravir

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Molnupiravir is a broad-spectrum antiviral prodrug of the nucleoside analog β-D-N4-hydroxycytidine (NHC). Molnupiravir competes predominantly with CTP for incorporation; it induces lethal viral mutagenesis by increasing G to A and C to U transition mutations without inhibition of RNA synthesis in replicating coronaviruses, which prevents the development of viral resistance. In primary airway epithelial cell cultures, Molnupiravir dose-dependently inhibits SARS-CoV-2 infectious virus production; it also inhibits MERS-CoV and SARS-CoV with EC50 values in the submicromolar range. In mice infected with SARS-CoV or MERS-CoV, Molnupiravir improves pulmonary function, and reduces virus titer and body weight loss. Molnupiravir also displays in vivo efficacy against seasonal and pandemic influenza A shed virus. It is orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | β-D-N4-hydroxycytidine-5 '-isopropyl ester |
| CAS | 2492423-29-5 |
| Quantity | 10 mg |
| Target | Antiviral Compounds |
| Molecular Formula | C13H19N3O7 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction