missing translation for 'onlineSavingsMsg'
Learn More

Molybdenite, naturally occurring mineral, grains, approximately 0.06-0.19in

Product Code. 11345357
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11345357 5 g 5g
11325357 1 g 1g
11335357 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11345357 Supplier Thermo Scientific Alfa Aesar Supplier No. 042642.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Used for grain mounts, XRD and microprobe standards, as well as identification of unknown minerals. | CAS: 1309-56-4 | MoS2 | 160.07 g/mol

Molybdenite, naturally occurring mineral, grains, is used for grain mounts, XRD and microprobe standards, as well as identification of unknown minerals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1309-56-4
Molecular Formula MoS2
Molecular Weight (g/mol) 160.07
MDL Number MFCD00003470
InChI Key CWQXQMHSOZUFJS-UHFFFAOYSA-N
Synonym molybdenum disulfide, molybdenite, molybdenum iv sulfide, molybdenum disulphide, molysulfide, molykote, motimol, nichimoly c, sumipowder pa, molykote z
PubChem CID 14823
ChEBI CHEBI:30704
IUPAC Name bis(sulfanylidene)molybdenum
SMILES S=[Mo]=S

Specifications

Quantity 5 g
Solubility Information Soluble in sulfuric and nitric acid
Formula Weight 160.07
Packaging Glass bottle
Chemical Name or Material Molybdenite

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.