missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum hexacarbonyl, 98%

Product Code. 10072490 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
Product Code. Quantity Packaging unitSize
10072490 250 g Glass bottle 250g
10496463 10 g Glass bottle 10g
10646022 50 g Glass bottle 50g
3 options
This item is not returnable. View return policy

Product Code. 10072490

Brand: Thermo Scientific Acros 190392500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Molybdenum hexacarbonyl, 98%, C6O6Mo, CAS Number-13939-06-5, hexacarbonylmolybdenu, carbon monooxide-molybdenum 6:1, hexacarbonylmolybdenum, may contain up to cr 5g, hexakis carbon monoxide molybdenum, molybdenumhexacarbonyl, hexakis methanidylidyneoxidanium molybdenum, 50g, 100.0, 97.5, CHEBI:30508 | CAS: 13939-06-5 | C6O6Mo | 264 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13939-06-5
Molecular Formula C6O6Mo
Molecular Weight (g/mol) 264
MDL Number MFCD00003466
InChI Key KMKBZNSIJQWHJA-UHFFFAOYSA-N
Synonym molybdenumhexacarbonyl, carbon monooxide-molybdenum 6:1, hexakis carbon monoxide molybdenum, hexacarbonylmolybdenu, hexakis methanidylidyneoxidanium molybdenum, hexacarbonylmolybdenum, may contain up to cr 5g
PubChem CID 2724068
ChEBI CHEBI:30508
IUPAC Name carbon monoxide;molybdenum
SMILES [C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Mo]

Specifications

Melting Point 150.0°C
Color White through Yellow
CAS Min % 97.5
CAS Max % 100.0
Physical Form Crystalline Powder
Quantity 250 g
Assay Percent Range 98%
Linear Formula Mo(CO)6
Fieser 02,287; 03,206; 04,346; 09,317; 10,274; 13,194; 01,140; 02,448; 03,325; 04,559; 05,736; 06,652; 07,68; 08,109; 09,113; 15,9
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in benzene, slightly soluble in ether and most oter common, organic solvents
Formula Weight 264
Percent Purity 98%
Infrared Spectrum Authentic
Packaging Glass bottle
Specific Gravity 1.96
Density 1.9600g/mL
Chemical Name or Material Molybdenum hexacarbonyl, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Molybdenum hexacarbonyl
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
Hazard Statement
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.