missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum plate, 4.76mm (0.188in) thick, 99.95% (metals basis)

Product Code. 15456435
Change view
Click to view available options
Quantity:
100 x 100 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15456435 100 x 100 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15456435 Supplier Thermo Scientific Alfa Aesar Supplier No. 042143.GH
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7439-98-7 | Mo | 95.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7439-98-7
Quantity 100 x 100 mm
Weight ≈486g/100x100mm
Form Plate
Molecular Formula Mo
MDL Number MFCD00003465
Synonym molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
InChI Key ZOKXTWBITQBERF-UHFFFAOYSA-N
SMILES [Mo]
IUPAC Name molybdenum
Molecular Weight (g/mol) 95.95
PubChem CID 23932
ChEBI CHEBI:28685
Percent Purity 99.95%
Assay (metals basis)
Chemical Name or Material Molybdenum plate, 4.76mm (0.188 in.) thick
Show More Show Less
compliance-icons
Product Identifier
  • Molybdenum foil
Signal Word
  • Warning
Hazard Category
  • Reproductive toxicity Category 2
Hazard Statement
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P403-Store in a well-ventilated place.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Molybdenum

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.