missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum silicide sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.5% (metals basis), Thermo Scientific™

Product Code. 15494835
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15494835 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15494835 Supplier Thermo Scientific Alfa Aesar Supplier No. 041076.KS
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Magnetron sputtering source

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Color Gray
CAS 12136-78-6
Melting Point 1870°C to 2030°C
Density 6.31 g/mL
Quantity 1 Ea.
Odor Odorless
Molecular Formula MoSi2
MDL Number MFCD00151535
Synonym molybdenum disilicide, molybdenum silicide, ms silicide, molybdenum-silicide, molybdenum silicide, mo5si3, molybdenum silicide sputtering target, 50.8mm 2.0in dia x 3.18mm 0.125in thick, molybdenum silicide sputtering target, 50.8mm 2.0in dia x 6.35mm 0.250in thick, molybdenum silicide sputtering target, 76.2mm 3.0in dia x 3.18mm 0.125in thick, molybdenum silicide sputtering target, 76.2mm 3.0in dia x 6.35mm 0.250in thick
InChI Key YXTPWUNVHCYOSP-UHFFFAOYSA-N
SMILES [Si]=[Mo]=[Si]
IUPAC Name bis($l^{2}-silanylidene)molybdenum
Molecular Weight (g/mol) 152.12
PubChem CID 6336985
Formula Weight 152.13
Percent Purity 99.5%
Assay (metals basis)
Chemical Name or Material Molybdenum silicide sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.