missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum(V) chloride, 95%, Thermo Scientific Chemicals

Catalog Number 10002550
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass bottle
10241-05-1
Cl5Mo
273.20
MFCD00003467
GICWIDZXWJGTCI-UHFFFAOYSA-I
molybdenum v chloride, molybdenum pentachloride, molybdenum chloride mocl5, mocl5, pentachloridomolybdenum, molybdenum 5+ chloride, acmc-1bx8l, ksc491a7h
61497
CHEBI:30635
[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Mo+5]
10241-05-1
Cl5Mo
273.20
MFCD00003467
GICWIDZXWJGTCI-UHFFFAOYSA-I
molybdenum v chloride, molybdenum pentachloride, molybdenum chloride mocl5, mocl5, pentachloridomolybdenum, molybdenum 5+ chloride, acmc-1bx8l, ksc491a7h
61497
CHEBI:30635
[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Mo+5]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY
Boiling Point 268°C
Melting Point 194°C
Color Black to Blue or Black-Green
CAS Min % 94.0
CAS Max % 100.0
Physical Form Fine Crystals or Crystalline Powder
Quantity 100 g
Assay Percent Range 94% min. (Argentometry)
Linear Formula MoCl5
UN Number 2508
Fieser 06,412; 01,1015; 02,365; 03,250; 04,428; 05,587; 06,519; 08,443; 09,414; 11,471; 12,434; 13,273; 04,99; 16,95
Solubility Information Solubility in water: reacts with water. Other solubilities: soluble in dry ether,dry alcohol and other,organic solvents
IUPAC Name pentachloromolybdenum
Formula Weight 273.2
Percent Purity 95%
Packaging Glass bottle
Chemical Name or Material Molybdenum(V) chloride, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Molybdenum(V) chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'