missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum(VI) Oxide, 99.5%, Solstice

Produktkod. 15686740
Change view
Klicka för att se tillgängliga alternativ
Quantity:
100 g
500 g
Packaging:
Poly bottle
Förpackningsstorlek:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity Packaging unitSize
15686740 500 g Poly bottle 500g
15676740 100 g Poly bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 15686740 Leverantör Honeywell-Fluka Leverantörsnummer 69850500G
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Currently Unavailable: We have no information when this product will be available.
Se alternativa produkter
Denna artikel kan inte returneras. Se returpolicy

Puriss. p.a., 99.5%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 1313-27-5
Molecular Formula MoO3
Molecular Weight (g/mol) 143.95
MDL Number MFCD00003469
InChI Key JKQOBWVOAYFWKG-UHFFFAOYSA-N
Synonym molybdenum trioxide, molybdenum vi oxide, molybdic anhydride, molybdic trioxide, molybdena, molybdic oxide, molybdenum oxide, molybdic acid anhydride, molybdenum peroxide
PubChem CID 14802
ChEBI CHEBI:30627
IUPAC Name trioxomolybdenum
SMILES O=[Mo](=O)=O

Specifikationer

Melting Point 795°c
Color White to Yellow
Physical Form Powder
Quantity 500 g
Linear Formula MoO3
UN Number NONH for all modes of transport
Formula Weight 143.94g/mol
Percent Purity 99.5%
Odor Odorless
Packaging Poly bottle
Density 4.70g/cm3
Chemical Name or Material Molybdenum(VI) oxide
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.