missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum(VI) oxide, ≥99.5%, Reagent Grade, Honeywell™

Product Code. 15657920 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Poly bottle
Unit Size:
100g
1kg
500g
This item is not returnable. View return policy

Product Code. 15657920

Brand: Honeywell Research Chemicals M0753100G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade,≥99.5%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-27-5
Molecular Formula MoO3
Molecular Weight (g/mol) 143.95
MDL Number MFCD00003469
InChI Key JKQOBWVOAYFWKG-UHFFFAOYSA-N
Synonym molybdenum trioxide, molybdenum vi oxide, molybdic anhydride, molybdic trioxide, molybdena, molybdic oxide, molybdenum oxide, molybdic acid anhydride, molybdenum peroxide
PubChem CID 14802
ChEBI CHEBI:30627
IUPAC Name trioxomolybdenum
SMILES O=[Mo](=O)=O

Specifications

Melting Point 795°C
Color Gray
Physical Form Powder
Quantity 100 g
Linear Formula MoO3
UN Number NONH for all modes of transport
Formula Weight 143.94g/mol
Percent Purity ≥99.5%
Odor Odorless
Packaging Poly bottle
Chemical Name or Material Molybdenum(VI) oxide
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.