missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum(VI) oxide, 99+%

Product Code. 10042670 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10042670 100 g Plastic bottle 100g
11994151 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10042670 Supplier Thermo Scientific Acros Supplier No. 206361000
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1313-27-5 | MoO3 | 143.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-27-5
Molecular Formula MoO3
Molecular Weight (g/mol) 143.95
MDL Number MFCD00003469
InChI Key JKQOBWVOAYFWKG-UHFFFAOYSA-N
Synonym molybdenum trioxide, molybdenum vi oxide, molybdic anhydride, molybdic trioxide, molybdena, molybdic oxide, molybdenum oxide, molybdic acid anhydride, molybdenum peroxide
PubChem CID 14802
ChEBI CHEBI:30627
IUPAC Name trioxomolybdenum
SMILES O=[Mo](=O)=O

Specifications

Boiling Point 1155.0°C
Melting Point 795.0°C
Color Green-Yellow
CAS Min % 99.0
CAS Max % 100.0
Physical Form Powder
Quantity 100 g
Packaging Plastic bottle
Assay Percent Range 99% min
Linear Formula MoO3
Fieser 04,203; 13,279; 16,227
Merck Index 15, 6325
Solubility Information Solubility in water: 0.5g/L (20°C). Other solubilities: soluble in conc. mineral acids, sol. of alkali, hydroxides and ammonia, soluble in potassium bitartrate
Formula Weight 143.94
Percent Purity ≥99%
Chemical Name or Material Molybdenum(VI) oxide, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Molybdenum(VI) oxide
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.