missing translation for 'onlineSavingsMsg'
Learn More

Molymod™ Nitrogen Component

Product Code. 12889455
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
12889455 10 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12889455 Supplier Molymod Supplier No. MA300
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Specifications

Color Blue
Diameter (Metric) 23 mm
For Use With (Equipment) For Molymod Models
Quantity 10 Pk.
Product Type Nitrogen Component
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.