missing translation for 'onlineSavingsMsg'
Learn More

Mometasone furoate, MedChemExpress

Product Code. 16451967
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16451967 50 mg 50mg
16451096 200 mg 200mg
16436286 100 mg 100mg
16421696 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16451967 Supplier Medchem Express Supplier No. HY13693
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Mometasone furoate (Sch32088) is a glucocorticoid receptor agonist with anti-inflammatory and anti-allergic activity. Mometasone furoate acts as a corticosteroid agent and used for topical applications in chronic skin eczema and airway inflammation management of asthma in vivo

Mometasone furoate (Sch32088) is a glucocorticoid receptor agonist with anti-inflammatory and anti-allergic activity. Mometasone furoate acts as a corticosteroid agent and used for topical applications in chronic skin eczema and airway inflammation management of asthma in vivo

Specifications

Chemical Name or Material Mometasone furoate
CAS 83919-23-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C27H30Cl2O6
Quantity 50 mg
Synonym Sch32088
Solubility Information DMSO : 50 mg/mL (95.89 mM; Need ultrasonic)
SMILES C[C@@]12[C@](C(CCl)=O)(OC(C3=CC=CO3)=O)[C@H](C)C[C@@]1([H])[C@]4([H])CCC5=CC(C=C[C@]5(C)[C@@]4(Cl)[C@@H](O)C2)=O
Molecular Weight (g/mol) 521.43
Formula Weight 521.43
Percent Purity 99.05%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.