missing translation for 'onlineSavingsMsg'
Learn More

Mono-benzyl malonate, 95%

Product Code. 11995021 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11995021 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11995021 Supplier Thermo Scientific Acros Supplier No. 439800100
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 40204-26-0 | C10H10O4 | 194.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 40204-26-0
CAS Min % 94.0
CAS Max % 100.0
Melting Point 45°C to 48°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Molecular Formula C10H10O4
MDL Number MFCD00138132
Quantity 10 g
Synonym mono-benzyl malonate, monobenzyl malonate, 3-benzyloxy-3-oxopropanoic acid, benzyl hemimalonate, benzyl hydrogen malonate, monobenzylmalonate, 2-benzyloxy carbonyl acetic acid, malonic acid monobenzyl ester, 3-benzyl-oxy-3-oxopropanoic acid, propanedioic acid, 1-phenylmethyl ester
InChI Key CFLAHQSWDKNWPW-UHFFFAOYSA-N
SMILES OC(=O)CC(=O)OCC1=CC=CC=C1
IUPAC Name 3-oxo-3-phenylmethoxypropanoic acid
Molecular Weight (g/mol) 194.19
PubChem CID 181659
ChEBI CHEBI:84093
Formula Weight 194.19
Percent Purity 95%
Physical Form Crystalline Powder or Powder
Chemical Name or Material Mono-benzyl malonate
Show More Show Less
compliance-icons
Product Identifier
  • mono-Benzyl malonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.