missing translation for 'onlineSavingsMsg'
Learn More

Mono(carboxymethyl) Phthalate, TRC

Product Code. 30480827
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30480827 1 g 1g
30468563 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30480827 Supplier Toronto Research Chemicals Supplier No. TRCM5256001G
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30435-30-4
Molecular Formula C10H8O6
Molecular Weight (g/mol) 224.17
Synonym 1,2-Benzenedicarboxylic Acid 1-(Carboxymethyl) Ester, 1,2-Benzenedicarboxylic Acid Mono(carboxymethyl) Ester, Phthalic Ecid monoester with Glycolic Acid, Glycolyl Hydrogen Phthalate
IUPAC Name 2-(carboxymethoxycarbonyl)benzoic acid
SMILES OC(=O)COC(=O)c1ccccc1C(=O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C10H8O6/c11-8(12)5-16-10(15)7-4-2-1-3-6(7)9(13)14/h1-4H,5H2,(H,11,12)(H,13,14)
Formula Weight 224.0321
Chemical Name or Material Mono(carboxymethyl) Phthalate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.