missing translation for 'onlineSavingsMsg'
Learn More

mono-Methyl succinate, 95%

Product Code. 10431392 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
25g
This item is not returnable. View return policy

Product Code. 10431392

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3878-55-5 | C5H8O4 | 132.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3878-55-5
Molecular Formula C5H8O4
Molecular Weight (g/mol) 132.11
MDL Number MFCD00002788
InChI Key JDRMYOQETPMYQX-UHFFFAOYSA-N
Synonym monomethyl succinate, succinic acid monomethyl ester, methyl hydrogen succinate, mono-methyl succinate, butanedioic acid, monomethyl ester, 3-carbomethoxypropanoic acid, succinic acid, monomethyl ester, unii-ya2v724s0a, mono-methyl hydrogen succinate, butanedioic acid monomethyl ester
PubChem CID 77487
ChEBI CHEBI:75146
IUPAC Name 4-methoxy-4-oxobutanoic acid
SMILES COC(=O)CCC(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 55°C to 59°C
Color White
Boiling Point 151°C (20.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula HO2CCH2CH2CO2CH3
Quantity 10 g
Beilstein 02, 608
Formula Weight 132.11
Percent Purity 95%
Physical Form Crystalline Chunks or Crystals
Chemical Name or Material mono-Methyl succinate, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.