missing translation for 'onlineSavingsMsg'
Learn More

mono-n-Dodecyl phosphate, tech. 90%

Product Code. 11322727
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11322727 5 g 5g
11332727 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11322727 Supplier Thermo Scientific Alfa Aesar Supplier No. L11941.06
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 2627-35-2 | C12H27O4P | 266.32 g/mol

mono-n-Dodecyl phosphate is a lysophosphatidic acid (LPA) receptor ligand which serves as a specific agonist at EDG-4 (LPA2) (EC50=700 nM) and antagonist at EDG-7

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
mono-n-Dodecyl phosphate is a lysophosphatidic acid (LPA) receptor ligand which serves as a specific agonist at EDG-4 (LPA2) (EC50=700 nM) and antagonist at EDG-7

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2627-35-2
Molecular Formula C12H27O4P
Molecular Weight (g/mol) 266.32
MDL Number MFCD00013946
InChI Key TVACALAUIQMRDF-UHFFFAOYSA-N
Synonym dodecyl phosphate, monolauryl phosphate, phosphoric acid, monododecyl ester, mono-n-dodecyl phosphate, monododecyl phosphate, dihydrogen monolauryl phosphate, lauryl dihydrogen phosphate, unii-0k08l8ge2h, phosphoric acid monododecyl ester, dodecyloxyphosphonic acid
PubChem CID 75816
IUPAC Name dodecyl dihydrogen phosphate
SMILES CCCCCCCCCCCCOP(O)(O)=O

Specifications

Melting Point 39°C to 45°C
Odor Odorless
Quantity 5 g
Beilstein 1710518
Solubility Information Insoluble in water.
Formula Weight 266.32
Percent Purity 90%
Grade Technical
Chemical Name or Material mono-n-Dodecyl phosphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.