missing translation for 'onlineSavingsMsg'
Learn More

Monobenzyl Phthalate, TRC

Product Code. 30471070
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30471070 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30471070 Supplier Toronto Research Chemicals Supplier No. TRCM524900100MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Monobenzyl Phthalate
CAS 2528-16-7
Purity Grade Notes HPLC
Molecular Formula C15 H12 O4
Quantity 100 mg
Synonym 1,2-Benzenedicarboxylic acid, mono(phenylmethyl) ester (9CI), Phthalic acid, benzyl ester (6CI,7CI,8CI), 2-[(Benzyloxy)carbonyl]benzoic acid, Benzyl hydrogen phthalate, Monobenzyl phthalate, NSC 402008
InChI Formula InChI=1S/C15H12O4/c16-14(17)12-8-4-5-9-13(12)15(18)19-10-11-6-2-1-3-7-11/h1-9H,10H2,(H,16,17)
SMILES OC(=O)c1ccccc1C(=O)OCc2ccccc2
IUPAC Name 2-phenylmethoxycarbonylbenzoic acid
Molecular Weight (g/mol) 256.25
Formula Weight 256.0736
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.