missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Monochlorobimane (mBCI)

Product Code. 11529086
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11529086 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11529086 Supplier Invitrogen™ Supplier No. M1381MP
Powering Proteomics
Use promo code 28003
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

monochlorobimane (mBCI)

Monochloromobimane is essentially nonfluorescent until conjugated, readily reacts with several low molecular weight thiols, including glutathione, N-acetylcysteine, mercaptopurine, peptides and plasma thiols. The glutathione conjugate of monochlorobimane has absorption/emission maxima ∼394/490 nm.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 25 mg
Product Type Monochlorobimane
Content And Storage Store in freezer -5°C to -30°C and protect from light.
Shipping Condition Room Temperature
Emission 490

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.