missing translation for 'onlineSavingsMsg'
Learn More

Monocrotaline N-Oxide, TRC

Product Code. 30010082
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010082 50 mg 50mg
30007499 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30010082 Supplier Toronto Research Chemicals Supplier No. TRCM52600550MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35337-98-5
Molecular Formula C16 H23 N O7
Molecular Weight (g/mol) 341.36
Synonym 2H-[1,6]Dioxacycloundecino[2,3,4-gh]pyrrolizine-2,6(3H)-dione, 4,5,8,10,12,13,13a,13b-octahydro-4,5-dihydroxy-3,4,5-trimethyl-, 11-oxide, (3R,4S,5S,13aR,13bR)- (9CI), 20-Norcrotalanan-11,15-dione, 14,19-dihydro-12,13-dihydroxy-, 4-oxide, (13α,14α)-, Monocrotaline, N-oxide (6CI,7CI), Monocrotaline oxide
SMILES C[C@H]1C(=O)O[C@@H]2CC[N+]3([O-])CC=C(COC(=O)[C@@](C)(O)[C@@]1(C)O)[C@H]23

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C16H23NO7/c1-9-13(18)24-11-5-7-17(22)6-4-10(12(11)17)8-23-14(19)16(3,21)15(9,2)20/h4,9,11-12,20-21H,5-8H2,1-3H3/t9-,11+,12+,15-,16+,17?/m0/s1
Formula Weight 341.1475
Chemical Name or Material Monocrotaline N-Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.