missing translation for 'onlineSavingsMsg'
Learn More

Monodecyl Phthalate, TRC

Product Code. 30466103
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30466103 1 g 1g
30482421 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30466103 Supplier Toronto Research Chemicals Supplier No. TRCM5267801G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24539-60-4
Molecular Formula C18H26O4
Molecular Weight (g/mol) 306.4
Synonym 1,2-Benzenedicarboxylic Acid 1-Decyl Ester, 1,2-Benzenedicarboxylic Acid, Monodecyl Ester, Phthalic Acid Monodecyl Ester, Decyl Hydrogen Phthalate, Monodecyl Phthalate
IUPAC Name 2-decoxycarbonylbenzoic acid
SMILES OC(C1=CC=CC=C1C(OCCCCCCCCCC)=O)=O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C18H26O4/c1-2-3-4-5-6-7-8-11-14-22-18(21)16-13-10-9-12-15(16)17(19)20/h9-10,12-13H,2-8,11,14H2,1H3,(H,19,20)
Formula Weight 306.1831
Chemical Name or Material Monodecyl Phthalate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.