missing translation for 'onlineSavingsMsg'
Learn More

Monoethanolamine, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Product Code. 15548574 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Narrow Mouth Amber Glass Bottle
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15548574 500 mL Narrow Mouth Amber Glass Bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15548574 Supplier Avantor J.T.Baker Supplier No. 931401
  • ONLINE EXCLUSIVE PRICE
    460.62 €
    341.62 € / 500mL
    Save 119.00 €
    25% Off
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 141-43-5
Molecular Formula C2H7NO
Molecular Weight (g/mol) 61.08
MDL Number MFCD00008183
InChI Key HZAXFHJVJLSVMW-UHFFFAOYSA-N
Synonym ethanolamine, monoethanolamine, aminoethanol, 2-hydroxyethylamine, colamine, glycinol, olamine, 2-amino-1-ethanol, ethanol, 2-amino, ethylolamine
PubChem CID 700
ChEBI CHEBI:16000
IUPAC Name 2-aminoethan-1-ol
SMILES NCCO

Specifications

Flash Point 86.00°C
Packaging Narrow Mouth Amber Glass Bottle
Quantity 500 mL
Specific Gravity 1L = 1.02kg
Formula Weight 61.08
Grade BAKER ANALYZED™ ACS Reagent
Chemical Name or Material Monoethanolamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.