missing translation for 'onlineSavingsMsg'
Learn More

Monoethyl Potassium Malonate, TRC

Product Code. 30470779
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30470779 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30470779 Supplier Toronto Research Chemicals Supplier No. TRCM54253010G
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Monoethyl Potassium Malonate
CAS 6148-64-7
Molecular Formula C5H7KO4
Quantity 10 g
Synonym Propanedioic Acid, 1-Ethyl Ester Potassium Salt (1:1), Malonic Acid Monoethyl Ester, Potassium Salt, Propanedioic Acid, Monoethyl Ester Potassium Salt, 3-Ethoxy-3-oxopropanoic Acid Potassium Salt, Ethyl Malonate Potassium Salt, Ethyl Potassium Malonate, Malonic Ethyl Ester Potassium Salt, Monoethyl Malonate Potassium Salt, Potassium Ethyl Malonate, Potassium Monoethyl Malonate
InChI Formula InChI=1S/C5H8O4.K/c1-2-9-5(8)3-4(6)7;/h2-3H2,1H3,(H,6,7);/q;+1/p-1
SMILES [K+].CCOC(=O)CC(=O)[O-]
IUPAC Name potassium;3-ethoxy-3-oxopropanoate
Molecular Weight (g/mol) 170.2
Formula Weight 169.9981
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.