missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monosodium Glutamate Monohydrate, TRC
Specifications
Specifications
| Chemical Name or Material | Monosodium Glutamate Monohydrate |
| CAS | 6106-04-3 |
| Molecular Formula | C5 H8 N O4 . H2 O . Na |
| Quantity | 25 g |
| Synonym | L-Glutamic acid, sodium salt, hydrate (1:1:1), Glutamic acid, monosodium salt, monohydrate, L- (8CI), L-Glutamic acid, monosodium salt, monohydrate (9CI), MSG monohydrate, Monosodium L-glutamate monohydrate, Monosodium glutamate monohydrate, Sodium glutamate monohydrate |
| InChI Formula | InChI=1S/C5H9NO4.Na.H2O/c6-3(5(9)10)1-2-4(7)8;;/h3H,1-2,6H2,(H,7,8)(H,9,10);;1H2/q;+1;/p-1/t3-;;/m0../s1 |
| SMILES | O.[Na+].N[C@@H](CCC(=O)O)C(=O)[O-] |
| IUPAC Name | sodium;(2S)-2-amino-5-hydroxy-5-oxopentanoate;hydrate |
| Molecular Weight (g/mol) | 187.126 |
| Formula Weight | 187.046 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?