missing translation for 'onlineSavingsMsg'
Learn More

Monosodium Glutamate Monohydrate, TRC

Product Code. 30475053
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
Product Code. Quantity unitSize
30475053 100 g 100g
1 options
This item is not returnable. View return policy

Product Code. 30475053

Brand: Toronto Research Chemicals TRCM567150100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Monosodium Glutamate Monohydrate
CAS 6106-04-3
Molecular Formula C5 H8 N O4 . H2 O . Na
Quantity 100 g
Synonym L-Glutamic acid, sodium salt, hydrate (1:1:1), Glutamic acid, monosodium salt, monohydrate, L- (8CI), L-Glutamic acid, monosodium salt, monohydrate (9CI), MSG monohydrate, Monosodium L-glutamate monohydrate, Monosodium glutamate monohydrate, Sodium glutamate monohydrate
InChI Formula InChI=1S/C5H9NO4.Na.H2O/c6-3(5(9)10)1-2-4(7)8;;/h3H,1-2,6H2,(H,7,8)(H,9,10);;1H2/q;+1;/p-1/t3-;;/m0../s1
SMILES O.[Na+].N[C@@H](CCC(=O)O)C(=O)[O-]
IUPAC Name sodium;(2S)-2-amino-5-hydroxy-5-oxopentanoate;hydrate
Molecular Weight (g/mol) 187.126
Formula Weight 187.046
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.