missing translation for 'onlineSavingsMsg'
Learn More

Pro-Lab Diagnostics™ Monovalent Somatic (O) Salmonella Antisera

Product Code. 11989054
Change view
Click to view available options
Grade:
Species: Group B, Antigen Factor 5
Species: Group B/D, Antigen Factors 12
Species: Group C, Antigen Factor 6 and 7
Species: Group E1, Antigen Factors 10
Species: Group E2, Antigen Factor 15
Species: Group E4, Antigen Factor 19
Species: Group G, Antigen Factors 13, 22 and 23
Species: Group G1, Antigen Factor 22
Species: Group G2, Antigen Factor 23
Species: Group K, Antigen Factor 18
Species: Group P, Antigen Factor 38
Unit Size:
3mL
Each
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Grade unitSize
11989054 Species: Group C, Antigen Factor 6 and 7 3mL
11919064 Species: Group B/D, Antigen Factors 12 Each
11949064 Species: Group E2, Antigen Factor 15 Each
11939064 Species: Group E1, Antigen Factors 10 Each
11959064 Species: Group E4, Antigen Factor 19 Each
11989064 Species: Group G, Antigen Factors 13, 22 and 23 Each
11999064 Species: Group G1, Antigen Factor 22 Each
11909074 Species: Group G2, Antigen Factor 23 Each
11949074 Species: Group K, Antigen Factor 18 Each
11979054 Species: Group B, Antigen Factor 5 Each
11999074 Species: Group P, Antigen Factor 38 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Product Code. 11989054 Supplier Pro-Lab Diagnostics™ Supplier No. PL.6013
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Use in slide agglutination testing for identification of Salmonella species/antigens according to Kauffmann-White classification. Pro-Lab Diagnostics™ Monovalent Somatic (O) Salmonella Antisera is a comprehensive range of monovalent Salmonella antisera delivered in economical 3.0mL bottles.

While polyvalent O (somatic) antisera aid in initial serogrouping, full identification of O antigens is achieved using monovalent-specific O antisera. The serotype of Salmonella isolates can then be determined using H (flagella) antisera.

  • Ready-to-use diluted antisera with 0.01% thimerosal as a preservative
  • Produced using reference strains prepared by World Health Organization guidelines
  • Certified reference center performance
  • Presented in color-coded dropper bottles
  • Long shelf life: up to 5 years from date of manufacture

Specifications

Certifications/Compliance W.H.O.
Content And Storage 2°C to 8°C, Do Not Freeze
Grade Species: Group C, Antigen Factor 6 and 7
Product Type Antiserum
For Use With (Application) Traditional Slide Agglutination Testing
Packaging Type Dropper Bottle
Quantity 3.0 mL
Slide Type Agglutination
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.