missing translation for 'onlineSavingsMsg'
Learn More

Montelukast Acyl-Beta-D-glucuronide-d6, TRC

Product Code. 30478039
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478039 10 mg 10mg
30481353 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30478039 Supplier Toronto Research Chemicals Supplier No. TRCM56801210MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C41 D6 H38 Cl N O9 S
Molecular Weight (g/mol) 768.344
Synonym 1-[1-[[[(1R)-1-[3-[(1E)-2-(7-Chloro-2-quinolinyl)ethenyl]phenyl]-3-[2-(1-hydroxy-1-methylethyl-d6)phenyl]propyl]thio]methyl]cyclopropaneacetate] beta-D-Glucopyranuronic Acid
IUPAC Name (2S,3S,4S,5R,6S)-6-[2-[1-[[(1R)-1-[3-[(E)-2-(7-chloroquinolin-2-yl)ethenyl]phenyl]-3-[2-(1,1,1,3,3,3-hexadeuterio-2-hydroxypropan-2-yl)phenyl]propyl]sulfanylmethyl]cyclopropyl]acetyl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid
SMILES [2H]C([2H])([2H])C(O)(c1ccccc1CC[C@@H](SCC2(CC(=O)O[C@@H]3O[C@@H]([C@@H](O)[C@H](O)[C@H]3O)C(=O)O)CC2)c4cccc(\C=C\c5ccc6ccc(Cl)cc6n5)c4)C([2H])([2H])[2H]

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C41H44ClNO9S/c1-40(2,50)30-9-4-3-7-25(30)13-17-32(27-8-5-6-24(20-27)10-15-29-16-12-26-11-14-28(42)21-31(26)43-29)53-23-41(18-19-41)22-33(44)51-39-36(47)34(45)35(46)37(52-39)38(48)49/h3-12,14-16,20-21,32,34-37,39,45-47,50H,13,17-19,22-23H2,1-2H3,(H,48,49)/b15-10+/t32-,34+,35+,36-,37+,39-/m1/s1/i1D3,2D3
Formula Weight 767.28
Chemical Name or Material Montelukast Acyl-beta-D-glucuronide-d6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.