missing translation for 'onlineSavingsMsg'
Learn More

Morpholine-4-carboxamidine hydrobromide, 98%

Product Code. 11438867
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11438867 5 g 5g
11448867 25 g 25g
11450421 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11438867 Supplier Thermo Scientific Alfa Aesar Supplier No. A19985.06
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 157415-17-3 | C5H12BrN3O | 210.075 g/mol

It is used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from oxidizing agents, water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 157415-17-3
Molecular Formula C5H12BrN3O
Molecular Weight (g/mol) 210.075
MDL Number MFCD00051994
InChI Key IOHSDHMCZXCESZ-UHFFFAOYSA-N
Synonym morpholine-4-carboximidamide hydrobromide, morpholinoformamidine hydrobromide, 4-morpholinylformamidine hydrobromide, morpholine-4-carboxamidine hydrobromide, 4-morpholinecarboximidamide, monohydrobromide, acmc-20adpt, morpholine-4-carboxamidine, bromide, morpholin-4-carboxamidine hydrobromide
PubChem CID 2806564
IUPAC Name morpholine-4-carboximidamide;hydrobromide
SMILES C1COCCN1C(=N)N.Br

Specifications

Melting Point ∼155°C to 165°C
Quantity 5 g
Beilstein 7389038
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 210.08
Percent Purity 98%
Chemical Name or Material Morpholine-4-carboxamidine hydrobromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.