missing translation for 'onlineSavingsMsg'
Learn More

Morpholine-4-carboximidamide hydrobromide, 98%, BLD Pharm™

Product Code. 30778221
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30778221 1 g 1g
30770207 5 g 5g
30777617 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30778221 Supplier BLD Pharm Supplier No. BD1404291g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 157415-17-3
Molecular Formula C5H12BrN3O
MDL Number MFCD00051994
Quantity 1 g
SMILES Br.NC(=N)N1CCOCC1
Molecular Weight (g/mol) 210.07
Percent Purity 98%
Chemical Name or Material Morpholine-4-carboximidamide hydrobromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.