missing translation for 'onlineSavingsMsg'
Learn More

Morpholine, 99.5%, purified by redistillation, AcroSeal™

Product Code. p-4813561 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Unit Size:
100mL
800mL
This item is not returnable. View return policy

Product Code. 11986831

Brand: Thermo Scientific Acros 433851000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 110-91-8 | C4H9NO | 87.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-91-8
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00005972
InChI Key YNAVUWVOSKDBBP-UHFFFAOYSA-N
Synonym 1-oxa-4-azacyclohexane, tetrahydro-1,4-oxazine, diethylene oximide, diethylenimide oxide, diethyleneimide oxide, drewamine, diethylene imidoxide, tetrahydro-p-oxazine, tetrahydro-2h-1,4-oxazine, p-isoxazine, tetrahydro
PubChem CID 8083
ChEBI CHEBI:34856
IUPAC Name morpholine
SMILES C1COCCN1

Specifications

Melting Point -5.0°C
Density 0.9900g/mL
Boiling Point 126.0°C to 130.0°C
Flash Point 32°C
Assay Percent Range 99.4% min. (GC)
Packaging AcroSeal™ Glass bottle
Refractive Index 1.4530 to 1.4550
Quantity 100 mL
Merck Index 15,6362
Specific Gravity 0.99
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol, ethanol, acetone, benzene,, ether, castor oil, ethylene glycol, 2-hexanone,, linseed oil, turpentine, pine oil
Viscosity 2.23 mPa.s (20°C)
Formula Weight 87.12
Percent Purity 99.5%
Chemical Name or Material Morpholine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Morpholine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.