missing translation for 'onlineSavingsMsg'
Learn More

Morpholine, 99+%, ACS reagent

Product Code. 10258853 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
2.5kg
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10258853 2.5 kg Glass bottle 2.5kg
10055710 5 g Glass bottle 5g
10209940 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Artikelnummer. 10258853 Leverandør Thermo Scientific Acros Leverandørnr. 415160025
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.

CAS: 110-91-8 | C4H9NO | 87.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 110-91-8
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00005972
InChI Key YNAVUWVOSKDBBP-UHFFFAOYSA-N
Synonym 1-oxa-4-azacyclohexane, tetrahydro-1,4-oxazine, diethylene oximide, diethylenimide oxide, diethyleneimide oxide, drewamine, diethylene imidoxide, tetrahydro-p-oxazine, tetrahydro-2h-1,4-oxazine, p-isoxazine, tetrahydro
PubChem CID 8083
ChEBI CHEBI:34856
IUPAC Name morpholine
SMILES C1COCCN1

Tekniske data

CAS Min % 99.0
CAS Max % 100.0
Melting Point -5.0°C
Density 0.9900g/mL
Boiling Point 126.0°C to 130.0°C
Flash Point 32°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Refractive Index 1.4530 to 1.4550
Quantity 2.5 kg
Beilstein 27, 5
Fieser 01,705; 11,352; 13,14
Merck Index 15, 6362
Specific Gravity 0.99
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol, ethanol, acetone, benzene,, ether, castor oil, ethylene glycol, 2-hexanone,, linseed oil, turpentine, pine oil
Viscosity 2.23 mPa.s (20°C)
Formula Weight 87.12
Vapor Pressure 11mbar at 20°C
Percent Purity 99+%
Grade ACS Reagent
Physical Form Liquid
Chemical Name or Material Morpholine, 99+%
Vis mere Vis mindre
compliance-icons compliance-icons compliance-icons
Produktidentifikator
  • Morpholine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.