missing translation for 'onlineSavingsMsg'
Learn More

Morroniside, MedChemExpress

Product Code. 16489008
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16489008 10 mg 10mg
16471428 5 mg 5mg
16463876 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16489008 Supplier Medchem Express Supplier No. HYN0532
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Morroniside has neuroprotective effect by inhibiting neuron apoptosis and MMP2/9 expression.

Morroniside has neuroprotective effect by inhibiting neuron apoptosis and MMP2/9 expression.

Specifications

Chemical Name or Material Morroniside
CAS 25406-64-8
Color White
Purity Grade Notes Research
Molecular Formula C17H26O11
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (246.08 mM; Need ultrasonic) ∣H2O : 50 mg/mL (123.04 mM; Need ultrasonic)
SMILES O[C@H]([C@@H](O)[C@@H]1O)[C@](O[C@@H]1CO)([H])O[C@H]2[C@@]([C@@H]3C)([H])[C@@](C[C@H](O)O3)([H])C(C(OC)=O)=CO2
Molecular Weight (g/mol) 406.38
Formula Weight 406.38
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.