missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Morusin

Product Code. 16433886
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16433886 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16433886 Supplier MedChemExpress Supplier No. HYN0622
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Morusin is a prenylated flavonoid isolated from M. australis with various biological activities, such as antitumor, antioxidant, and anti-bacteria property. Morusin could inhibit NF-κB and STAT3 activity.

Specifications

Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 420.45 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-Kinase/protease
CAS 62596-29-6
Product Type Natural Products
Purity 99.83%
Solubility DMSO : ≥ 100 mg/mL (237.84 mM)
Synonym Mulberrochromene
Target NF-κB∣STAT∣Bacterial
Quantity 25 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.