missing translation for 'onlineSavingsMsg'
Learn More

uvex Motion S1 SRC Perforated Safety Shoes

Product Code. 15378312 Shop All Uvex Products
Change view
Click to view available options
Size:
40
41
43
44
Color:
Black, Silver
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Color Unit Size
15378312 44 Black, Silver Each
15338312 40 Black, Silver Each
15348312 41 Black, Silver Each
15368312 43 Black, Silver Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15378312 Supplier uvex Supplier No. 6989844
  • ONLINE EXCLUSIVE PRICE
    130.00 €
    121.71 € / Pack of 1
    Save 8.29 €
    6% Off
View all promotions
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extremely lightweight safety shoes with a sporty design constructed from synthetic materials, making them suitable for those who are allergic to chrome. Protection class S1 in accordance with EN ISO 20345:2011 with additional marking for excellent slip resistance (SRC).

  • Lightweight S1 safety shoes with a Climazone climate management technology
  • Made from synthetic materials, making them suitable for those who are allergic to chrome
  • Protection class S1 in accordance with EN ISO 20345:2011, additional marking for excellent slip resistance (SRC)
  • Meets the ESD specification with bleeder resistor under 35 megaohms
  • Self-cleaning, lightweight PU outsole with optimal abrasion resistance, shock absorption, and excellent slip resistance
  • Steel toe cap with a stabilizing anti-twist heel cap to prevent ankle twisting
  • All soles are free of silicones, plasticizers, and other paint wetting impairment substances
  • Anti-static Uvex 3D Hydroflex™ foam insole for optimal shock absorption around heel and forefoot
  • Elastic laces allow for quick, individual adjustment
  • Suitable for use with orthopaedic adjustments and insoles
TRUSTED_SUSTAINABILITY

Specifications

Color Black, Silver
Liner Material Distance Mesh
Material Microvelour
Size 44
Sole Material Dual Density Polyurethane (PU)
Toe Material Steel
Width (Metric) 11 cm
Gender Men's
Resistant To Oil, petrol
Model Motion S1 SRC
Ankle Style Trainers
Waterproof No
EN ISO 20345 S1
Product Type ESD Safety Shoe
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.