missing translation for 'onlineSavingsMsg'
Learn More

Berthold Technologies Mount For Test Tube in LB 9509

Product Code. 31777292 Shop All Berthold Technologies Products
Change view
Click to view available options
Color:
Black
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
31777292 Black Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31777292 Supplier Berthold Technologies Supplier No. 57011
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For luminescence test tube in the Junior portable luminometer

  • Mount is required to calibrate the Junior portable luminometer using the Luminescence Test Tube LB 9517
  • Luminescence Test Tube LB 9517 is used to assess performance of the Junior luminometer

Specifications

Type Mount
Color Black
For Use With (Equipment) Junior LB 9509 Portable Luminometer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.