missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse anti-Bovine IgG (H+L) Secondary Antibody, HRP

Product Code. 15257544
Change view
Click to view available options
Quantity:
250 μg
Unit Size:
250µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15257544 250 μg 250µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15257544 Supplier Invitrogen™ Supplier No. MA516733
  • ONLINE EXCLUSIVE PRICE
    578.00 €
    514.42 € / 250µg
    Save 63.58 €
    11% Off
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Mouse Monoclonal Secondary Antibody

Mouse anti Bovine IgG, clone IL-A2 is specific for bovine Immunoglobulin G (IgG). The specificity of this antibody for bovine IgG1 and IgG2 has been demonstrated by ELISA.

Anti-Bovine secondary antibodies are affinity-purified antibodies with well-characterized specificity for bovine (cow, ox) immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bovine IgG (H+L)
Applications ELISA, Western Blot
Classification Monoclonal
Clone IL-A2
Concentration 1 mg/mL
Conjugate HRP
Formulation PBS with 0.01% thimerosal
Host Species Mouse
Immunogen Purified bovine IgG.
Purification Method Protein A
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Bovine
Content And Storage 4°C, do not freeze
Product Type Secondary Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.