missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse anti-Feline IgE Secondary Antibody
Mouse Monoclonal Secondary Antibody
Brand: Invitrogen™ SA510297
This item is not returnable.
View return policy
Description
Mouse anti-Feline IgE Secondary Antibody for Western Blot, ELISA
Anti-Feline secondary antibodies are affinity-purified antibodies with well-characterized specificity for feline immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
| Feline IgE | |
| Monoclonal | |
| 3.73 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Feline IgE. | |
| 1 mg | |
| Secondary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Western Blot | |
| ME1 | |
| Unconjugated | |
| Mouse | |
| Antigen affinity chromatography | |
| RUO | |
| Feline | |
| Secondary Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction