Learn More
Mouse anti-Hamster IgG2, Clone: G192-3, Secondary Antibody, BD
Brand: BD Biosciences 554029
Description
Based on ELISA, the G192-3 antibody reacts specifically with Syrian hamster IgG2 monoclonal antibodies. The G192-3 mAb does not react with other hamster IgG groups or hamster IgM.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature
Host Species: Mouse
Clone: G192-3
Isotype: IgG1, κ
Species Reactivity: Hamster
Immunogen: Pooled Armenian and Syrian Hamster IgG mAb
ELISA, Flow Cytometry
Specifications
| Mouse anti-Hamster IgG2 | |
| G192-3 | |
| Biotin | |
| 0.5 mg | |
| Immunology | |
| Hamster |
| Monoclonal | |
| 0.5mg/mL | |
| Mouse | |
| RUO | |
| Secondary | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.