Learn More
Mouse anti-Human IgD, Clone: IA6-2, Secondary Antibody, BD
Description
Reacts with the heavy chain of human IgD expressed on B cells. This reactivity is neutralized by purified human IgD and not by purified human IgM. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: IA6-2
Isotype: IgG2a, κ
Species Reactivity: Human
Flow Cytometry
Specifications
Specifications
| Antigen | Mouse anti-Human IgD |
| Classification | Monoclonal |
| Clone | IA6-2 |
| Concentration | 0.5mg/mL |
| Host Species | Mouse |
| Purification Method | Affinity Purified |
| Quantity | 0.5 mg |
| Regulatory Status | RUO |
| Research Discipline | Immunology |
| Primary or Secondary | Secondary |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.