missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse anti-Human IgE Secondary Antibody
Mouse Monoclonal Secondary Antibody
Brand: Invitrogen™ MII0204PG
This item is not returnable.
View return policy
Description
MII0204PG was formerly sold as a Seradyn product. This is a larger pack size of MA5-14727.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
| Human IgE | |
| Monoclonal | |
| 5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Human Myeloma IgE. | |
| 1 mg | |
| Secondary | |
| Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C | |
| Liquid |
| Western Blot, ELISA, Radioimmune Assays (RIA), Immunoprecipitation | |
| Fc-2001 | |
| Unconjugated | |
| Mouse | |
| Protein A | |
| RUO | |
| Human | |
| Secondary Antibody | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction