Learn More
Mouse anti-Human IgG, Clone: G18-145, Secondary Antibody, BD
Description
Reacts with the heavy chain of human immunoglobulin subclasses: IgG1, IgG2, IgG3 and IgG4. Does not react with other heavy chain isotypes. This antibody is routinely tested by ELISA analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: G18-145
Isotype: IgG1, κ
Species Reactivity: Human
ELISA, Flow Cytometry
Specifications
Specifications
| Antigen | Mouse anti-Human IgG |
| Classification | Monoclonal |
| Clone | G18-145 |
| Concentration | 0.5mg/mL |
| Host Species | Mouse |
| Purification Method | Affinity Purified |
| Quantity | 0.5 mg |
| Regulatory Status | RUO |
| Research Discipline | Immunology |
| Primary or Secondary | Secondary |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.