missing translation for 'onlineSavingsMsg'
Learn More

Mouse Anti-Human IgG, APC, Clone: G18-145, Secondary Antibody, BD

Brand:  BD Biosciences 550931

Product Code. 15806438

  • ONLINE EXCLUSIVE PRICE
    401.00 €
    384.13 € / Each
    Save 16.87 €
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reacts with the heavy chain of human immunoglobulin subclasses: IgG1, IgG2, IgG3 and IgG4. Does not react with other heavy chain isotypes.

Host Species: Mouse
Clone: G18-145
Isotype: IgG1 κ
Species Reactivity: Human

ELISA, Flow Cytometry

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Mouse Anti-Human IgG
G18-145
Mouse
RUO
Secondary
IgG1 κ
Monoclonal
APC
100 Tests
Immunology
Human
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.